| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009185.hg.1 |
ENST00000351396.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009185.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009185.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009185.hg.1 |
TC10000772.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160024.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160024.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160024.hg.1 |
ENST00000369839.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160024.hg.1 |
NM_006951.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160024.hg.1 |
TC1000008769.hg.1 |
Clariom D TC |