Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00908892_m1
          See other FOXP1 GE Assays ›
          Gene Symbol:
          FOXP1
          Gene Name:
          forkhead box P1
          Gene Aliases:
          12CC4, HSPC215, MFH, QRF1, hFKH1B
          Chromosome Location:
          Chr.3: 70954714 - 71583989 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00908892_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FOXP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001244808.1 NP_001231737.1 19 - 20 2349 80
          NM_001244810.1 NP_001231739.1 19 - 20 2297 80
          NM_001244812.1 NP_001231741.1 14 - 15 1649 80
          NM_001244813.1 NP_001231742.1 13 - 14 1541 80
          NM_001244814.1 NP_001231743.1 15 - 16 2046 80
          NM_001244815.1 NP_001231744.1 14 - 15 1987 80
          NM_001244816.1 NP_001231745.1 18 - 19 2154 80
          NM_032682.5 NP_116071.2 19 - 20 2249 80
          XM_005264735.3 XP_005264792.1 14 - 15 1935 80
          XM_005264736.3 XP_005264793.1 14 - 15 1984 80
          XM_005264737.4 XP_005264794.1 19 - 20 2255 80
          XM_005264742.3 XP_005264799.1 13 - 14 1541 80
          XM_006713102.2 XP_006713165.1 18 - 19 2210 80
          XM_006713103.2 XP_006713166.1 16 - 17 1947 80
          XM_006713104.2 XP_006713167.1 17 - 18 2180 80
          XM_011533584.2 XP_011531886.1 19 - 20 2320 80
          XM_011533585.2 XP_011531887.1 18 - 19 2172 80
          XM_011533588.2 XP_011531890.1 14 - 15 1770 80
          XM_017006165.1 XP_016861654.1 18 - 19 2091 80
          XM_017006166.1 XP_016861655.1 18 - 19 2187 80
          XM_017006167.1 XP_016861656.1 14 - 15 1981 80
          GenBank mRNA AB052767.1 - 9 - 10 986 80
          AF146696.1 - 17 - 18 1986 80
          AF151049.1 - 2 - 3 141 80
          AF250920.1 - 18 - 19 2154 80
          AF275309.1 - 10 - 11 1094 80
          AK027264.1 - 14 - 15 1619 80
          AK074321.1 - 15 - 16 2045 80
          AK074569.1 - 13 - 14 1542 80
          AK092383.1 - 14 - 15 1649 80
          AK122710.1 - 19 - 20 2349 80
          AK297649.1 - 14 - 15 1452 80
          CR457164.1 - 1 - 2 49 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300011485.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001546.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FOXP1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of B cell apoptotic process
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          positive regulation of endothelial cell migration
          osteoclast differentiation
          response to lipopolysaccharide
          regulation of tumor necrosis factor production
          chemokine (C-C motif) ligand 2 secretion
          osteoclast development
          macrophage activation
          monocyte activation
          endothelial cell activation
          regulation of monocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of smooth muscle cell proliferation
          regulation of interleukin-1 beta secretion
          regulation of inflammatory response
          negative regulation of androgen receptor signaling pathway
          T follicular helper cell differentiation
          interleukin-21 secretion
          regulation of defense response to bacterium
          regulation of macrophage colony-stimulating factor production
          regulation of endothelial tube morphogenesis
          regulation of interleukin-12 secretion
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          protein binding
          sequence-specific DNA binding
          protein self-association
          metal ion binding
          androgen receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000318789.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000318796.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000358280.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000468577.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000475937.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000484350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000491238.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000493089.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 ENST00000498215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_001244808.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_001244810.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_001244812.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_001244813.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_001244814.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_001244815.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_001244816.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 NM_032682.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028179.hg.1 TC03001546.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000318789.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000327590.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000460805.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000468577.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000475937.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000484350.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000491238.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000493089.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000497355.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000498215.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000614176.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000615603.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244808.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244810.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244812.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244814.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244815.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244816.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_032682.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000318789.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000327590.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000460805.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000468577.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000475937.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000484350.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000491238.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000493089.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000497355.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000498215.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000614176.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 ENST00000615603.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244808.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244810.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244812.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244814.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244815.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_001244816.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 NM_032682.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300186058.hg.1 TC0300011485.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline