Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00908958_g1
          See other SORBS1 GE Assays ›
          Gene Symbol:
          SORBS1
          Gene Name:
          sorbin and SH3 domain containing 1
          Gene Aliases:
          CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
          Chromosome Location:
          Chr.10: 95311773 - 95561371 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00908958_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SORBS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034954.3 NP_001030126.2 7 - 8 515 60
          NM_001034955.2 NP_001030127.2 5 - 6 377 60
          NM_001034956.2 NP_001030128.2 5 - 6 377 60
          NM_001290294.2 NP_001277223.2 7 - 8 515 60
          NM_001290295.2 NP_001277224.2 6 - 7 419 60
          NM_001290297.3 NP_001277226.2 6 - 7 419 60
          NM_001377198.1 NP_001364127.1 6 - 7 419 60
          NM_001377199.1 NP_001364128.1 7 - 8 515 60
          NM_001377200.1 NP_001364129.1 6 - 7 419 60
          NM_001377203.1 NP_001364132.1 7 - 8 515 60
          NM_001377204.1 NP_001364133.1 6 - 7 419 60
          NM_001377206.1 NP_001364135.1 7 - 8 515 60
          NM_001377207.1 NP_001364136.1 6 - 7 419 60
          NM_001377208.1 NP_001364137.1 7 - 8 515 60
          NM_001377209.1 NP_001364138.1 6 - 7 419 60
          NM_001384447.1 NP_001371376.1 6 - 7 419 60
          NM_001384449.1 NP_001371378.1 6 - 7 419 60
          NM_001384450.1 NP_001371379.1 6 - 7 419 60
          NM_001384451.1 NP_001371380.1 6 - 7 419 60
          NM_001384452.1 NP_001371381.1 6 - 7 419 60
          NM_001384453.1 NP_001371382.1 6 - 7 419 60
          NM_001384454.1 NP_001371383.1 7 - 8 541 60
          NM_001384455.1 NP_001371384.1 5 - 6 333 60
          NM_001384456.1 NP_001371385.1 7 - 8 670 60
          NM_001384457.1 NP_001371386.1 7 - 8 541 60
          NM_001384458.1 NP_001371387.1 6 - 7 419 60
          NM_001384459.1 NP_001371388.1 7 - 8 515 60
          NM_001384460.1 NP_001371389.1 7 - 8 515 60
          NM_001384461.1 NP_001371390.1 6 - 7 419 60
          NM_001384463.1 NP_001371392.1 8 - 9 637 60
          NM_001384464.1 NP_001371393.1 6 - 7 419 60
          NM_001419685.1 NP_001406614.1 7 - 8 515 60
          NM_001419702.1 NP_001406631.1 7 - 8 515 60
          NM_001419703.1 NP_001406632.1 8 - 9 621 60
          NM_001419704.1 NP_001406633.1 7 - 8 515 60
          NM_015385.4 NP_056200.2 6 - 7 419 60
          NM_024991.3 NP_079267.2 7 - 8 515 60
          XM_011539150.3 XP_011537452.1 7 - 8 515 60
          XM_017015501.3 XP_016870990.1 7 - 8 515 60
          XM_017015502.3 XP_016870991.1 7 - 8 515 60
          XM_017015503.3 XP_016870992.1 7 - 8 515 60
          XM_017015505.3 XP_016870994.1 7 - 8 515 60
          XM_017015507.3 XP_016870996.1 7 - 8 515 60
          XM_017015510.3 XP_016870999.1 7 - 8 515 60
          XM_017015515.3 XP_016871004.1 7 - 8 515 60
          XM_017015517.3 XP_016871006.1 7 - 8 515 60
          XM_017015523.2 XP_016871012.1 6 - 7 419 60
          XM_017015525.2 XP_016871014.1 6 - 7 419 60
          XM_017015530.3 XP_016871019.1 7 - 8 515 60
          XM_017015536.2 XP_016871025.1 6 - 7 419 60
          XM_047424468.1 XP_047280424.1 7 - 8 515 60
          XM_047424469.1 XP_047280425.1 7 - 8 515 60
          XM_047424472.1 XP_047280428.1 7 - 8 515 60
          XM_047424473.1 XP_047280429.1 6 - 7 419 60
          XM_047424477.1 XP_047280433.1 7 - 8 515 60
          XM_047424479.1 XP_047280435.1 6 - 7 419 60
          GenBank mRNA AB037717.1 - 7 - 8 496 60
          AF330623.1 - 3 - 4 236 60
          AF356525.1 - 5 - 6 388 60
          AF356526.1 - 5 - 6 388 60
          AF356527.1 - 5 - 6 388 60
          AK022468.1 - 7 - 8 543 60
          AK300407.1 - 6 - 7 421 60
          AL117472.1 - 6 - 7 426 60
          AM260536.1 - 7 - 8 522 60
          BC152463.1 - 7 - 8 496 60

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SORBS1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          insulin receptor signaling pathway
          cell-substrate adhesion
          cellular response to insulin stimulus
          stress fiber assembly
          positive regulation of glycogen biosynthetic process
          positive regulation of glucose import
          positive regulation of insulin receptor signaling pathway
          positive regulation of lipid biosynthetic process
          focal adhesion assembly
          positive regulation of protein localization to plasma membrane
          actin binding
          insulin receptor binding
          protein binding
          cytoskeletal protein binding
          receptor signaling complex scaffold activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline