Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00909325_s1
          See other CDKN2A-AS1 GE Assays ›
          Gene Symbol:
          CDKN2A-AS1
          Gene Name:
          CDKN2A antisense RNA 1 (head to head)
          Gene Aliases:
          C9orf53, bA149I2.3
          Chromosome Location:
          Chr.9: 21967139 - 21967754 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00909325_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDKN2A-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024274.1 1 146 60
          GenBank mRNA AF211119.1 - 1 164 60
          BX091351.1 - 1 146 60

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26928991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26928993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26928992 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 74973 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 43246 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P307502 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P307502 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 33480 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9535 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P307502 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P307502 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26928999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640775 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26928997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26928998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640775 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26929009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06640749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26928999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26928997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26928998 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P307502 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P121053 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P307502 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640772 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16165517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640827 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640825 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640814 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640754 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640773 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640830 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640762 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640747 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640820 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640805 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640767 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640816 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640760 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16165503 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640765 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640777 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640808 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640757 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640834 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640744 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640826 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640763 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640756 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640752 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640775 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640823 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640811 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640741 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640831 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640818 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640829 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640750 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06640770 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P121053 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8154655 - -
          Affymetrix Human Gene 1.1 ST Array 8154654 - -
          Affymetrix HT HG-U133+ PM Array Plate 220505_PM_at - -
          Affymetrix Human Genome U219 Array 11744769_at - -
          Affymetrix Human Genome U219 Array 11744769_PM_at - -
          Human Genome U133 Plus 2.0 Array 220505_PM_at - -
          Human Genome U133 Plus 2.0 Array 220505_at - -
          Human Genome U133A 2.0 Array 220505_at - -
          Human Genome U133A 2.0 Array 220505_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw528284 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220505_at11 - -
          Human Genome U95 Set 73929_PM_at - -
          Human Genome U95 Set 73929_at - -
          Human Genome U133 Set 220505_PM_at - -
          Human Genome U133 Set 220505_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038427 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038427 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 116479 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1687924 NR_024274.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1687924 AF211119.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1687924 BX091351.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1687924 NR_024274.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1687924 AF211119.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1687924 BX091351.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1687924 NR_024274.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1687924 AF211119.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1687924 BX091351.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1687924 NR_024274.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1687924 AF211119.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1687924 BX091351.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1687924 NR_024274.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1687924 AF211119.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1687924 BX091351.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline