Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00911143_g1
          See other ALOX12 GE Assays ›
          Gene Symbol:
          ALOX12
          Gene Name:
          arachidonate 12-lipoxygenase, 12S type
          Gene Aliases:
          12-LOX, 12S-LOX, LOG12
          Chromosome Location:
          Chr.17: 6993791 - 7010736 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00911143_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ALOX12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000697.2 NP_000688.2 1 - 2 189 108
          XM_011523780.2 XP_011522082.1 1 - 2 2463 108
          GenBank mRNA BC069557.1 - 1 - 2 167 7262463 108
          M35418.1 - 1 - 2 177 108
          M58704.1 - 1 - 2 169 108
          M62982.1 - 1 - 2 189 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700006717.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000081.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ALOX12

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          aging
          positive regulation of cell proliferation
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of muscle cell apoptotic process
          arachidonic acid metabolic process
          lipoxygenase pathway
          fatty acid oxidation
          positive regulation of cell growth
          positive regulation of cell migration
          superoxide anion generation
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          linoleic acid metabolic process
          positive regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of vasodilation
          positive regulation of smooth muscle cell proliferation
          hepoxilin metabolic process
          hepoxilin biosynthetic process
          positive regulation of mitochondrial depolarization
          establishment of skin barrier
          cellular response to lipid
          reactive oxygen species metabolic process
          negative regulation of platelet aggregation
          leukotriene A4 metabolic process
          lipoxin metabolic process
          lipoxin A4 biosynthetic process
          lipoxin B4 biosynthetic process
          arachidonate 12-lipoxygenase activity
          iron ion binding
          protein binding
          linoleate 13S-lipoxygenase activity
          oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
          hepoxilin-epoxide hydrolase activity
          hepoxilin A3 synthase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009608.hg.1 ENST00000573222.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009608.hg.1 ENST00000573222.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17000986.hg.1 ENST00000251535.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17000986.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17000986.hg.1 TC17000081.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17000985.hg.1 ENST00000251535.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17000985.hg.1 NM_000697.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17000985.hg.1 TC17000081.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000406228.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 ENST00000480801.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006717.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700146370.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700146370.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700146370.hg.1 TC1700006717.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700146370.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700146370.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay TC1700009608.hg.1 ENST00000573222.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009608.hg.1 ENST00000573222.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700146371.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700146371.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700146371.hg.1 TC1700006717.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700146371.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700146371.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700146369.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700146369.hg.1 NM_000697.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700146369.hg.1 TC1700006717.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700146369.hg.1 ENST00000251535.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700146369.hg.1 NM_000697.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline