Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00915304_m1
          See other TECPR2 GE Assays ›
          Gene Symbol:
          TECPR2
          Gene Name:
          tectonin beta-propeller repeat containing 2
          Gene Aliases:
          KIAA0329, SPG49
          Chromosome Location:
          Chr.14: 102362912 - 102502481 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00915304_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TECPR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001172631.2 NP_001166102.1 1 - 2 205 119
          NM_014844.4 NP_055659.2 1 - 2 205 119
          GenBank mRNA AB002327.1 - 1 - 2 154 119
          BC151230.1 - 1 - 2 178 8860116 119
          BP365224.1 - 1 - 2 207 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008349.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000781.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AB002327.1,BC151230.1,NM_001172631.2,NM_014844.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TECPR2

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000359520.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000391028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000557786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000557786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000557786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000557786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000557786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000558678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000559124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000559124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000559124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000559124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000559124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000560060.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000560060.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000560060.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000560060.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000560060.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 ENST00000561228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_001172631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008349.hg.1 NM_014844.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline