Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00916518_m1
          See other SP1 GE Assays ›
          Gene Symbol:
          SP1
          Gene Name:
          Sp1 transcription factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 53380195 - 53416446 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00916518_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001251825.1 NP_001238754.1 1 - 2 106 122
          NM_138473.2 NP_612482.2 1 - 2 106 122
          GenBank mRNA BC062539.1 - 1 - 2 107 5928633 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007713.hg
          This assay targets the same location as HTA transcript cluster(s) TC12000437.hg

          Additional Information:

          This assay is in the 5'UTR region of the BC062539.1,NM_001251825.1,NM_138473.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SP1

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          viral process
          snRNA transcription from RNA polymerase II promoter
          positive regulation by host of viral transcription
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          regulation of phenotypic switching by transcription from RNA polymerase II promoter
          positive regulation of hydrogen sulfide biosynthetic process
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          core promoter sequence-specific DNA binding
          RNA polymerase II repressing transcription factor binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein C-terminus binding
          transcription factor binding
          histone acetyltransferase binding
          protein homodimerization activity
          histone deacetylase binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding
          HMG box domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12005667.hg.1 ENST00000327443.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12005667.hg.1 ENST00000551969.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12005667.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12005667.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12005667.hg.1 TC12000437.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12005670.hg.1 ENST00000327443.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12005670.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12005670.hg.1 ENST00000551969.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12005670.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12005670.hg.1 NM_003109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12005670.hg.1 NM_138473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12005670.hg.1 TC12000437.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200155225.hg.1 TC1200007713.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200155226.hg.1 TC1200007713.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200155233.hg.1 TC1200007713.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000327443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000426431.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000548560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 ENST00000551969.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_001251825.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_003109.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007713.hg.1 NM_138473.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline