Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00916666_m1
          See other TSTD2 GE Assays ›
          Gene Symbol:
          TSTD2
          Gene Name:
          thiosulfate sulfurtransferase like domain containing 2
          Gene Aliases:
          C9orf97
          Chromosome Location:
          Chr.9: 97600080 - 97633680 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00916666_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TSTD2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139246.4 NP_640339.4 3 - 4 974 73
          XM_011518312.2 XP_011516614.1 2 - 3 437 73
          GenBank mRNA AF258575.1 - 2 - 3 420 73
          AK056453.1 - 1 - 2 101 73
          AK094043.1 - 3 - 4 687 73
          AK122809.1 - 1 - 2 1088 73
          AK128615.1 - 4 - 5 640 73
          AK291199.1 - 3 - 4 595 73
          BC035604.1 - 3 - 4 621 4385905 73
          BX538308.1 - 3 - 4 615 73
          BX648493.1 - 3 - 4 613 73
          KU178877.1 - 2 - 3 405 73
          KU178878.1 - 2 - 3 377 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010946.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001395.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF258575.1,AK056453.1,AK122809.1,XM_011518312.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TSTD2

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09017368.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017368.hg.1 ENST00000354801.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017368.hg.1 ENST00000375163.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017368.hg.1 ENST00000375165.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017368.hg.1 ENST00000375172.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017368.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017368.hg.1 TC09001395.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 ENST00000354801.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 ENST00000375163.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 ENST00000375165.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 ENST00000375172.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017366.hg.1 TC09001395.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174603.hg.1 TC0900010946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000375173.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010946.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 ENST00000484708.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174601.hg.1 TC0900010946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000341170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000375163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000375165.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 ENST00000375172.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 NM_139246.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900174602.hg.1 TC0900010946.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline