| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158647.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158647.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158647.hg.1 |
TC2000008318.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158647.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158647.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
TC2000008318.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158642.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158646.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158646.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158646.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158646.hg.1 |
TC2000008318.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158646.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158646.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158646.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |