Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00921511_g1
          See other AMPD1 GE Assays ›
          Gene Symbol:
          AMPD1
          Gene Name:
          adenosine monophosphate deaminase 1
          Gene Aliases:
          MAD, MADA, MMDD
          Chromosome Location:
          Chr.1: 114673098 - 114695618 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00921511_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AMPD1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000036.2 NP_000027.2 9 - 10 1372 107
          NM_001172626.1 NP_001166097.1 8 - 9 1360 107
          GenBank mRNA AK097077.1 - 8 - 9 1359 106
          AK291349.1 - 9 - 10 1276 107
          AK314252.1 - 9 - 10 1282 107
          M60092.1 - 9 - 10 1309 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015350.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003013.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AMPD1

          Panther Classification:

          Molecular Function -

          deaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to organic substance
          IMP salvage
          purine-containing compound salvage
          AMP deaminase activity
          myosin heavy chain binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01046992.hg.1 ENST00000353928.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01046992.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01046992.hg.1 ENST00000520113.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01046992.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01046992.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01046992.hg.1 TC01003013.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000485564.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000485564.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000485564.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000485564.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000485564.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000485564.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015350.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 ENST00000369538.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 ENST00000520113.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 NM_000036.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 NM_001172626.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214690.hg.1 TC0100015350.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline