Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00922008_g1
          See other FANCG GE Assays ›
          Gene Symbol:
          FANCG
          Gene Name:
          Fanconi anemia complementation group G
          Gene Aliases:
          FAG, XRCC9
          Chromosome Location:
          Chr.9: 35073838 - 35080016 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00922008_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FANCG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004629.1 NP_004620.1 7 - 8 1418 96
          GenBank mRNA AJ007669.1 - 7 - 8 1418 96
          AK293427.1 - 8 - 9 1108 96
          AK311348.1 - 6 - 7 892 96
          AK312987.1 - 7 - 8 1343 96
          BC000032.2 - 7 - 8 1064 3507258 96
          BC011623.2 - 7 - 8 1191 4053144 96
          BT009813.1 - 7 - 8 926 96
          KU159422.1 - 7 - 8 1449 96
          KU178053.1 - 7 - 8 926 96
          KU178054.1 - 7 - 8 926 96
          KU178055.1 - 7 - 8 926 96
          KU178056.1 - 4 - 5 500 96
          U70310.1 - 7 - 8 1060 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900009930.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001049.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FANCG

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle checkpoint
          ovarian follicle development
          DNA repair
          mitochondrion organization
          spermatid development
          response to radiation
          interstrand cross-link repair
          damaged DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000378643.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000425676.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000448890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000461149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000461149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000461149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000462124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000462124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000462124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000462124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000462124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000462124.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000474894.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000474894.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000474894.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000474894.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000476212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000476212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000476212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000476212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000481254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000481254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000481254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000481254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 ENST00000481254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009930.hg.1 NM_004629.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline