| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006122.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006122.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006122.hg.1 |
TC04000432.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006119.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006119.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006119.hg.1 |
TC04000432.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006123.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006123.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006123.hg.1 |
TC04000432.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154384.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154384.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154384.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154384.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154384.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154384.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154384.hg.1 |
TC0400007918.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400154382.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154382.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154382.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154382.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154382.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154382.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154382.hg.1 |
TC0400007918.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400154383.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154383.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154383.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154383.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154383.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154383.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154383.hg.1 |
TC0400007918.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154379.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154379.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154379.hg.1 |
ENST00000286758.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154379.hg.1 |
NM_006419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154379.hg.1 |
TC0400007918.hg.1 |
Clariom D TC |