Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00922332_m1
          See other SEPT8 GE Assays ›
          Gene Symbol:
          SEPT8
          Gene Name:
          septin 8
          Gene Aliases:
          SEP2
          Chromosome Location:
          Chr.5: 132750817 - 132778216 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00922332_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SEPT8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001098811.1 NP_001092281.1 2 - 3 393 63
          NM_001098812.1 NP_001092282.1 2 - 3 393 63
          NM_001098813.1 NP_001092283.1 2 - 3 319 63
          NM_001300798.1 NP_001287727.1 2 - 3 393 63
          NM_001300799.1 NP_001287728.1 2 - 3 393 63
          NM_015146.1 NP_055961.1 2 - 3 393 63
          XM_005271927.2 XP_005271984.1 2 - 3 395 63
          XM_005271928.2 XP_005271985.1 2 - 3 395 63
          XM_005271931.3 XP_005271988.1 3 - 4 476 63
          XM_011543288.1 XP_011541590.1 2 - 3 415 63
          XM_017009254.1 XP_016864743.1 2 - 3 395 63
          XM_017009255.1 XP_016864744.1 2 - 3 462 63
          XM_017009256.1 XP_016864745.1 2 - 3 462 63
          XM_017009257.1 XP_016864746.1 2 - 3 415 63
          XM_017009258.1 XP_016864747.1 2 - 3 462 63
          XM_017009259.1 XP_016864748.1 2 - 3 462 63
          XM_017009260.1 XP_016864749.1 2 - 3 415 63
          XM_017009261.1 XP_016864750.1 2 - 3 462 63
          GenBank mRNA AF179995.1 - 2 - 3 393 63
          AF440761.1 - 2 - 3 393 63
          AF440762.1 - 2 - 3 393 63
          AF440763.1 - 2 - 3 228 63
          AK057797.1 - 2 - 3 257 63
          BC140759.1 - 2 - 3 215 9021627 63
          BC171779.1 - 2 - 3 215 9054433 63
          D86957.1 - 2 - 3 393 63
          DA261871.1 - 2 - 3 462 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500012025.hg
          This assay targets the same location as HTA transcript cluster(s) TC05001772.hg

          Additional Information:

          This assay is in the 5'UTR region of the NM_001098813.1,XM_005271931.3,XM_011543288.1,XM_017009255.1,XM_017009256.1,XM_017009257.1,XM_017009258.1,XM_017009259.1,XM_017009260.1,XM_017009261.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Process(es)

          protein binding
          GTP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000296873.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000378699.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000378701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000378706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000378719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000378721.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000448933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 ENST00000458488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 NM_001098811.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 NM_001098812.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 NM_015146.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024044.hg.1 TC05001772.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05024060.hg.1 ENST00000378699.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024060.hg.1 ENST00000448933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024060.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024060.hg.1 TC05001772.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000296873.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000378699.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000378701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000378706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000378719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000378721.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000448933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 ENST00000458488.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 NM_001098811.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 NM_001098812.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 NM_015146.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024043.hg.1 TC05001772.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05024059.hg.1 ENST00000378699.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024059.hg.1 ENST00000448933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024059.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024059.hg.1 TC05001772.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500183098.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183098.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183098.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183098.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183098.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183098.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183098.hg.1 TC0500012025.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 ENST00000414594.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 ENST00000414594.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183099.hg.1 TC0500012025.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000296873.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000371478.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000371493.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378701.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378706.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378719.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378721.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000414594.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000453480.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000458488.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000481794.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000620483.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001098811.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001098812.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001300798.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001300799.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_015146.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000296873.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000371478.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000371493.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378701.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378706.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378719.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000378721.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000414594.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000453480.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000458488.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000481794.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 ENST00000620483.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001098811.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001098812.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001300798.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_001300799.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 NM_015146.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183081.hg.1 TC0500012025.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000296873.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000371478.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000371493.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378701.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378706.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378719.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378721.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000414594.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000453480.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000458488.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000481794.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000492490.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000620483.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001098811.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001098812.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001300798.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001300799.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_015146.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000296873.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000371478.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000371493.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378699.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378701.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378706.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378719.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000378721.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000414594.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000448933.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000453480.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000458488.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000481794.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000492490.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 ENST00000620483.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001098811.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001098812.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001098813.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001300798.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_001300799.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 NM_015146.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183082.hg.1 TC0500012025.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline