Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00922358_m1
          See other HIF1AN GE Assays ›
          Gene Symbol:
          HIF1AN
          Gene Name:
          hypoxia inducible factor 1 alpha subunit inhibitor
          Gene Aliases:
          FIH1
          Chromosome Location:
          Chr.10: 100535884 - 100553924 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00922358_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HIF1AN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017902.2 NP_060372.2 3 - 4 654 115
          XM_011539940.2 XP_011538242.1 3 - 4 665 115
          XM_011539941.1 XP_011538243.1 3 - 4 532 115
          GenBank mRNA AF395830.1 - 3 - 4 589 115
          AK000622.1 - 3 - 4 599 115
          AK295353.1 - 3 - 4 681 115
          AL359615.1 - 2 - 3 317 115
          BC007719.2 - 3 - 4 582 4138066 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008651.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000724.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HIF1AN

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          peptidyl-histidine hydroxylation
          peptidyl-aspartic acid hydroxylation
          peptidyl-asparagine hydroxylation
          positive regulation of myoblast differentiation
          negative regulation of Notch signaling pathway
          oxidation-reduction process
          regulation of transcription from RNA polymerase II promoter in response to hypoxia
          negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
          positive regulation of vasculogenesis
          Notch binding
          iron ion binding
          protein binding
          zinc ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
          oxygen sensor activity
          carboxylic acid binding
          peptidyl-histidine dioxygenase activity
          peptidyl-asparagine 3-dioxygenase activity
          protein homodimerization activity
          cofactor binding
          NF-kappaB binding
          ankyrin repeat binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10008428.hg.1 ENST00000299163.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008428.hg.1 ENST00000442724.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008428.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10008428.hg.1 TC10000724.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10008427.hg.1 ENST00000299163.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008427.hg.1 ENST00000442724.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008427.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10008427.hg.1 TC10000724.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000528044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008651.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000159002.hg.1 TC1000008651.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000299163.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000478787.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000526476.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 ENST00000533589.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 NM_017902.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000159003.hg.1 TC1000008651.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline