Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00923193_m1
          See other PPM1F GE Assays ›
          Gene Symbol:
          PPM1F
          Gene Name:
          protein phosphatase, Mg2+/Mn2+ dependent 1F
          Gene Aliases:
          CAMKP, CaMKPase, FEM-2, POPX2, hFEM-2
          Chromosome Location:
          Chr.22: 21919419 - 21952878 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00923193_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPM1F
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014634.3 NP_055449.1 5 - 6 896 87
          XM_011530556.1 XP_011528858.1 4 - 5 627 87
          GenBank mRNA AF305840.1 - 4 - 5 859 87
          AF520615.1 - 4 - 5 749 87
          AK126377.1 - 3 - 4 1306 87
          AK291628.1 - 5 - 6 866 87
          AK294557.1 - 3 - 4 753 87
          BC071989.1 - 5 - 6 838 5421075 87
          BC105599.1 - 5 - 6 862 6450839 87
          D13640.1 - 5 - 6 855 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200008110.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000548.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PPM1F

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of cell-substrate adhesion
          histone dephosphorylation
          negative regulation of peptidyl-serine phosphorylation
          cellular response to drug
          peptidyl-threonine dephosphorylation
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          negative regulation of transcription, DNA-templated
          positive regulation of growth
          positive regulation of chemotaxis
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          intrinsic apoptotic signaling pathway
          protein serine/threonine phosphatase activity
          protein binding
          phosphatase activity
          calmodulin-dependent protein phosphatase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22009914.hg.1 ENST00000263212.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009914.hg.1 ENST00000397495.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009914.hg.1 ENST00000406981.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009914.hg.1 ENST00000407142.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009914.hg.1 ENST00000538191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009914.hg.1 NM_014634.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009914.hg.1 TC22000548.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22009915.hg.1 ENST00000263212.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009915.hg.1 ENST00000397495.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009915.hg.1 ENST00000406981.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009915.hg.1 ENST00000407142.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009915.hg.1 ENST00000538191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009915.hg.1 NM_014634.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009915.hg.1 TC22000548.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22009911.hg.1 ENST00000263212.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009911.hg.1 ENST00000397495.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009911.hg.1 ENST00000406981.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009911.hg.1 ENST00000407142.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009911.hg.1 ENST00000538191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009911.hg.1 NM_014634.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009911.hg.1 TC22000548.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000263212.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000397495.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000407142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000445205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 NM_014634.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 TC2200008110.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000263212.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000397495.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000407142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 ENST00000445205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158029.hg.1 NM_014634.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000263212.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000397495.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000407142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000484588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 NM_014634.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 TC2200008110.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000263212.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000397495.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000407142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 ENST00000484588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158027.hg.1 NM_014634.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000263212.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000397495.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000407142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000445205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000623652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 NM_014634.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 TC2200008110.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000263212.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000397495.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000407142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000445205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 ENST00000623652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158031.hg.1 NM_014634.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline