Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00924395_m1
          See other CDK10 GE Assays ›
          Gene Symbol:
          CDK10
          Gene Name:
          cyclin dependent kinase 10
          Gene Aliases:
          PISSLRE
          Chromosome Location:
          Chr.16: 89686612 - 89696364 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00924395_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDK10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001098533.2 NP_001092003.2 6 - 7 488 65
          NM_001160367.1 NP_001153839.1 6 - 7 488 65
          NM_052987.3 NP_443713.2 6 - 7 488 65
          NM_052988.4 NP_443714.3 6 - 7 526 65
          NR_027702.1 5 - 6 453 65
          NR_027703.1 5 - 6 453 65
          XM_006721308.2 XP_006721371.1 6 - 7 582 65
          XM_006721310.2 XP_006721373.1 6 - 7 582 65
          XM_011523405.2 XP_011521707.1 6 - 7 582 65
          XM_011523406.2 XP_011521708.1 6 - 7 582 65
          XM_011523407.2 XP_011521709.1 6 - 7 582 65
          XM_011523408.2 XP_011521710.1 6 - 7 582 65
          XM_011523410.2 XP_011521712.1 6 - 7 500 65
          XM_011523411.2 XP_011521713.1 7 - 8 738 65
          XM_011523412.1 XP_011521714.1 7 - 8 708 65
          XM_011523413.2 XP_011521715.1 6 - 7 1140 65
          XM_011523414.2 XP_011521716.1 6 - 7 926 65
          XM_011523415.2 XP_011521717.1 5 - 6 457 65
          XM_011523416.2 XP_011521718.1 6 - 7 665 65
          XM_011523417.2 XP_011521719.1 6 - 7 582 65
          XM_017023806.1 XP_016879295.1 5 - 6 471 65
          XM_017023807.1 XP_016879296.1 6 - 7 582 65
          XM_017023808.1 XP_016879297.1 7 - 8 737 65
          XM_017023809.1 XP_016879298.1 5 - 6 457 65
          XM_017023810.1 XP_016879299.1 6 - 7 925 65
          GenBank mRNA AB209829.1 - 5 - 6 433 65
          AK131531.1 - 5 - 6 2085 65
          AK290485.1 - 6 - 7 446 65
          AK292351.1 - 6 - 7 504 65
          AK295397.1 - 6 - 7 492 65
          AK296631.1 - 5 - 6 430 65
          AK298386.1 - 6 - 7 526 65
          BC017342.2 - 6 - 7 468 4865641 65
          BC025301.1 - 5 - 6 421 4637860 65
          BC093028.1 - 6 - 7 505 65
          CX867290.1 - 6 - 7 491 65
          DB118903.1 - 5 - 6 453 65
          DC362478.1 - 6 - 7 526 65
          L33264.1 - 6 - 7 487 65
          X78342.1 - 6 - 7 603 65

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDK10

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          traversing start control point of mitotic cell cycle
          negative regulation of cell proliferation
          positive regulation of MAPK cascade
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline