Search
Search

PCDHA6| Gene Symbol: PCDHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018909.3 | NP_061732.1 | 1 - 2 | 2563 | 81 | |
| GenBank mRNA | AB209205.1 | - | 1 - 2 | 1339 | 81 | |
| AF152314.1 | - | 1 - 2 | 2393 | 81 | ||
| BC036674.2 | - | 1 - 2 | 2507 | 5263226 | 81 | |
Set Membership: |
|