Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00927127_m1
          See other UTS2 GE Assays ›
          Gene Symbol:
          UTS2
          Gene Name:
          urotensin 2
          Gene Aliases:
          PRO1068, U-II, UCN2, UII
          Chromosome Location:
          Chr.1: 7846345 - 7913615 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00927127_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UTS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017000116.1 XP_016855605.1 6 - 7 896 91
          XM_017000117.1 XP_016855606.1 5 - 6 541 91
          XM_017000118.1 XP_016855607.1 4 - 5 348 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          negative regulation of glomerular filtration
          muscle contraction
          positive regulation of cytosolic calcium ion concentration
          chemical synaptic transmission
          regulation of blood pressure
          negative regulation of heart rate
          positive regulation of heart rate
          positive regulation of fibroblast migration
          positive regulation of circadian sleep/wake cycle, wakefulness
          positive regulation of synaptic transmission, cholinergic
          positive regulation of collagen biosynthetic process
          response to testosterone
          negative regulation of urine volume
          negative regulation of renal sodium excretion
          response to drug
          positive regulation of cell differentiation
          positive regulation of angiogenesis
          negative regulation of blood pressure
          positive regulation of blood pressure
          positive regulation of vasodilation
          positive regulation of circadian sleep/wake cycle, REM sleep
          negative regulation of insulin secretion
          positive regulation of fibroblast proliferation
          receptor binding
          hormone activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000054668.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000361696.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 ENST00000377516.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_006786.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_021995.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012732.hg.1 NM_021995.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline