Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00928208_m1
          See other LINC01346 GE Assays ›
          Gene Symbol:
          LINC01346
          Gene Name:
          long intergenic non-protein coding RNA 1346
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 3940612 - 3952583 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00928208_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01346
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040065.1 2 - 3 240 68
          GenBank mRNA AK124708.1 - 2 - 3 235 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100006631.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000076.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15016012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15016010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021393 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15016017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15016016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021403 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021358 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021390 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15016009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15016008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021398 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021360 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20021361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00012592 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 81376 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 266210 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 502980 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 524733 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 425572 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P932339 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P932339 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 10424 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 13103 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P932339 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P932339 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P932339 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P00004032 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00004032 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 56736 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 69238 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 75282 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00004045 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00004032 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00004040 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00004045 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00004032 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1566737_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1566735_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01001221.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01001221.hg.1 NR_040065.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01001221.hg.1 TC01000076.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 NR_040065.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 NR_040065.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006631.hg.1 NR_040065.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100145923.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100145923.hg.1 NR_040065.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100145923.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100145923.hg.1 NR_040065.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100145923.hg.1 TC0100006631.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000412674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 ENST00000456897.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 NR_040065.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 NR_040065.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006631.hg.1 NR_040065.1 Refseq
          Human Genome U133 Plus 2.0 Array 1566737_PM_at - -
          Human Genome U133 Plus 2.0 Array 1566737_at - -
          Human Genome U133 Plus 2.0 Array 1566735_at - -
          Human Genome U133 Plus 2.0 Array 1566735_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566737_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566735_at10 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3228056 DB288420.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3228056 NR_040065.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3228056 AK124708.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3233656 DB288420.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline