| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023959.hg.1 |
ENST00000257749.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023959.hg.1 |
ENST00000343122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023959.hg.1 |
ENST00000537989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023959.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023959.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023959.hg.1 |
TC06001946.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008786.hg.1 |
ENST00000413986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008786.hg.1 |
ENST00000413986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000481150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000481150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000493201.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000493201.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000493201.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000406998.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000453877.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000470301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000472023.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000481150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000481150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000493201.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000493201.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000493201.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000494747.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012554.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008786.hg.1 |
ENST00000413986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008786.hg.1 |
ENST00000413986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008786.hg.1 |
ENST00000445838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
ENST00000257749.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
ENST00000343122.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
ENST00000537989.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
NM_001170794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
NM_021813.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186628.hg.1 |
TC0600012554.hg.1 |
Clariom D TC |