Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: HACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321080.1 | NP_001308009.1 | 18 - 19 | 2353 | 88 | |
NM_001321083.1 | NP_001308012.1 | 19 - 20 | 2300 | 88 | ||
NM_001321084.1 | NP_001308013.1 | 18 - 19 | 2217 | 88 | ||
NM_020771.3 | NP_065822.2 | 19 - 20 | 2485 | 88 | ||
NR_104424.1 | 21 - 22 | 2684 | 88 | |||
XM_017011113.1 | XP_016866602.1 | 19 - 20 | 2352 | 88 | ||
XM_017011114.1 | XP_016866603.1 | 18 - 19 | 2171 | 88 | ||
XM_017011115.1 | XP_016866604.1 | 18 - 19 | 2327 | 88 | ||
XM_017011116.1 | XP_016866605.1 | 18 - 19 | 2382 | 88 | ||
XM_017011118.1 | XP_016866607.1 | 16 - 17 | 1905 | 88 | ||
XM_017011119.1 | XP_016866608.1 | 18 - 19 | 2188 | 88 | ||
XM_017011120.1 | XP_016866609.1 | 17 - 18 | 2303 | 88 | ||
XM_017011121.1 | XP_016866610.1 | 17 - 18 | 2270 | 88 | ||
XM_017011122.1 | XP_016866611.1 | 18 - 19 | 2022 | 88 | ||
GenBank mRNA | AB037741.1 | - | 8 - 9 | 3232 | 88 | |
AK291760.1 | - | 19 - 20 | 2410 | 88 | ||
AK292683.1 | - | 8 - 9 | 999 | 88 | ||
AK294164.1 | - | 7 - 8 | 750 | 88 | ||
AK300314.1 | - | 8 - 9 | 926 | 88 | ||
AK316091.1 | - | 6 - 7 | 712 | 88 | ||
AL834202.1 | - | 6 - 7 | 722 | 88 | ||
BC034982.2 | - | 19 - 20 | 2449 | 4838835 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |