| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021490.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021490.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021490.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021490.hg.1 |
TC01001369.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021491.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021491.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021491.hg.1 |
TC01001369.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100175621.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175621.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175621.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175621.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175621.hg.1 |
TC0100010301.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100175620.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175620.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175620.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175620.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175620.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175620.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175620.hg.1 |
TC0100010301.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000368061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
ENST00000483408.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010301.hg.1 |
NM_020335.2 |
Refseq |