Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00937328_m1
          See other PDGFD GE Assays ›
          Gene Symbol:
          PDGFD
          Gene Name:
          platelet derived growth factor D
          Gene Aliases:
          IEGF, MSTP036, SCDGF-B, SCDGFB
          Chromosome Location:
          Chr.11: 103907186 - 104164299 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00937328_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDGFD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033135.3 NP_149126.1 1 - 2 496 86
          GenBank mRNA AY027518.1 - 1 - 2 216 86
          AY359116.1 - 1 - 2 155 86
          BC030645.2 - 1 - 2 576 4824526 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100012155.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11002242.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDGFD

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          blood coagulation
          positive regulation of cell proliferation
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of cell migration
          positive regulation of protein autophosphorylation
          cellular response to platelet-derived growth factor stimulus
          positive regulation of MAP kinase activity
          platelet-derived growth factor receptor signaling pathway
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of cell division
          cellular response to hydrogen peroxide
          positive regulation of ERK1 and ERK2 cascade
          cellular response to amino acid stimulus
          cellular response to transforming growth factor beta stimulus
          positive regulation of smooth muscle cell chemotaxis
          positive regulation of glomerular mesangial cell proliferation
          positive regulation of monocyte extravasation
          platelet-derived growth factor receptor binding
          growth factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11026799.hg.1 ENST00000302251.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026799.hg.1 ENST00000393158.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026799.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026799.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026799.hg.1 TC11002242.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11026800.hg.1 ENST00000302251.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026800.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026800.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026800.hg.1 TC11002242.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026797.hg.1 ENST00000302251.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026797.hg.1 ENST00000393158.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026797.hg.1 NM_025208.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026797.hg.1 NM_033135.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026797.hg.1 TC11002242.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100193431.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193431.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193431.hg.1 TC1100012155.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100193430.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193430.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193430.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193430.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193430.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193430.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193430.hg.1 TC1100012155.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193425.hg.1 TC1100012155.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193428.hg.1 TC1100012155.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 ENST00000302251.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 ENST00000393158.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 NM_025208.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 NM_033135.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100193429.hg.1 TC1100012155.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline