| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012789.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012789.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012789.hg.1 |
TC20000928.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009392.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165723.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165723.hg.1 |
NM_004776.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165723.hg.1 |
TC2000009392.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000165723.hg.1 |
ENST00000371711.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165723.hg.1 |
NM_004776.3 |
Refseq |