Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00943350_g1
          See other GSTP1 GE Assays ›
          Gene Symbol:
          GSTP1
          Gene Name:
          glutathione S-transferase pi 1
          Gene Aliases:
          DFN7, FAEES3, GST3, GSTP, HEL-S-22, PI
          Chromosome Location:
          Chr.11: 67583595 - 67586653 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00943350_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GSTP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000852.3 NP_000843.1 3 - 4 391 67
          GenBank mRNA BC010915.1 - 3 - 4 178 3960700 67
          BT019949.1 - 2 - 3 142 67
          BT019950.1 - 2 - 3 142 67
          CA946272.1 - 3 - 4 391 67
          CR450361.1 - 2 - 3 142 67
          EU794620.1 - 3 - 4 171 67
          U30897.1 - 3 - 4 154 67
          U62589.1 - 3 - 4 155 67
          X06547.1 - 2 - 3 148 67
          X15480.1 - 3 - 4 159 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100008144.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000694.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GSTP1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          negative regulation of acute inflammatory response
          negative regulation of protein kinase activity
          glutathione metabolic process
          xenobiotic metabolic process
          central nervous system development
          negative regulation of biosynthetic process
          negative regulation of tumor necrosis factor-mediated signaling pathway
          oligodendrocyte development
          organ regeneration
          response to estradiol
          negative regulation of interleukin-1 beta production
          negative regulation of tumor necrosis factor production
          cellular response to insulin stimulus
          regulation of stress-activated MAPK cascade
          negative regulation of stress-activated MAPK cascade
          positive regulation of superoxide anion generation
          response to L-ascorbic acid
          common myeloid progenitor cell proliferation
          nitric oxide storage
          negative regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to amino acid
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          negative regulation of JUN kinase activity
          linoleic acid metabolic process
          response to ethanol
          response to mercury ion
          negative regulation of fibroblast proliferation
          negative regulation of nitric-oxide synthase biosynthetic process
          regulation of ERK1 and ERK2 cascade
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of leukocyte proliferation
          cellular response to lipopolysaccharide
          cellular response to epidermal growth factor stimulus
          cellular response to glucocorticoid stimulus
          cellular response to cell-matrix adhesion
          negative regulation of monocyte chemotactic protein-1 production
          cellular oxidant detoxification
          glutathione derivative biosynthetic process
          negative regulation of extrinsic apoptotic signaling pathway
          glutathione transferase activity
          glutathione peroxidase activity
          protein binding
          drug binding
          JUN kinase binding
          kinase regulator activity
          S-nitrosoglutathione binding
          dinitrosyl-iron complex binding
          glutathione binding
          nitric oxide binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000495996.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000495996.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000495996.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009027.hg.1 ENST00000398603.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009027.hg.1 ENST00000398606.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009027.hg.1 ENST00000498765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009027.hg.1 NM_000852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009027.hg.1 TC11000694.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100159999.hg.1 TC1100008144.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398603.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000398606.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000467591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000489040.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000494593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000495996.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000495996.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000495996.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 ENST00000498765.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008144.hg.1 NM_000852.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 214807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0