Search
Search

MLLT10| Gene Symbol: MLLT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195626.1 | NP_001182555.1 | 11 - 12 | 1661 | 124 | |
| NM_001324297.1 | NP_001311226.1 | 14 - 15 | 2494 | 124 | ||
| NM_004641.3 | NP_004632.1 | 12 - 13 | 2010 | 172 | ||
| NR_136736.1 | 13 - 14 | 2188 | 242 | |||
| GenBank mRNA | AB209755.1 | - | 13 - 14 | 1894 | 242 | |
| AY598745.1 | - | 11 - 12 | 1661 | 124 | ||
| BC129946.1 | - | 12 - 13 | 1704 | 124 | ||
| BX648210.1 | - | 5 - 6 | 746 | 124 | ||
| U13948.1 | - | 12 - 13 | 1844 | 172 | ||
Set Membership: |
|