| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009456.hg.1 |
ENST00000575506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009456.hg.1 |
ENST00000575506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009456.hg.1 |
ENST00000575506.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009455.hg.1 |
ENST00000614400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009456.hg.1 |
ENST00000575506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009456.hg.1 |
ENST00000575506.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009456.hg.1 |
ENST00000575506.2 |
Ensembl |