| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019393.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019393.hg.1 |
ENST00000395833.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019393.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019393.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019393.hg.1 |
TC11001543.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019386.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019386.hg.1 |
ENST00000395833.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019386.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019386.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019386.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019386.hg.1 |
TC11001543.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019388.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019388.hg.1 |
ENST00000395833.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019388.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019388.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019388.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019388.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019388.hg.1 |
TC11001543.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179670.hg.1 |
TC1100010495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179674.hg.1 |
TC1100010495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000464025.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
ENST00000493667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010495.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000257818.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000395833.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000411482.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
ENST00000465614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
NM_001142315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
NM_001142316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
NM_005574.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100179668.hg.1 |
TC1100010495.hg.1 |
Clariom D TC |