| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007285.hg.1 |
ENST00000338437.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007285.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007285.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007285.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007285.hg.1 |
TC16000522.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
NM_005182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
TC1600008134.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
ENST00000338437.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
ENST00000394069.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
ENST00000548332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
NM_001014435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159967.hg.1 |
NM_005182.2 |
Refseq |