Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00952263_g1
          See other WNK4 GE Assays ›
          Gene Symbol:
          WNK4
          Gene Name:
          WNK lysine deficient protein kinase 4
          Gene Aliases:
          PHA2B, PRKWNK4
          Chromosome Location:
          Chr.17: 42779701 - 42797066 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00952263_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WNK4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321299.1 NP_001308228.1 6 - 7 1724 105
          NM_032387.4 NP_115763.2 7 - 8 1813 105
          XM_011525135.2 XP_011523437.1 7 - 8 2743 105
          XM_017024962.1 XP_016880451.1 7 - 8 2743 105
          XM_017024963.1 XP_016880452.1 7 - 8 2743 105
          XM_017024964.1 XP_016880453.1 7 - 8 2743 105
          XM_017024965.1 XP_016880454.1 7 - 8 2743 105
          XM_017024966.1 XP_016880455.1 4 - 5 865 105
          GenBank mRNA AF390018.1 - 7 - 8 1766 105
          AJ316534.1 - 7 - 8 1762 105
          AK096003.1 - 1 - 2 236 105
          AK096052.1 - 5 - 6 1282 105
          BC136664.1 - 7 - 8 1763 9020654 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007915.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000536.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene WNK4

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          ion transport
          chloride transport
          protein localization
          positive regulation of ion transmembrane transporter activity
          intracellular signal transduction
          regulation of cellular process
          ion homeostasis
          renal sodium ion absorption
          distal tubule morphogenesis
          negative regulation of pancreatic juice secretion
          regulation of ion homeostasis
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          chloride channel inhibitor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17006517.hg.1 ENST00000246914.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006517.hg.1 ENST00000316085.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006517.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006517.hg.1 TC17000536.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000587745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000587745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 ENST00000592669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000587705.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000587745.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000587745.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 ENST00000592669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007915.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700157726.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700157726.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700157726.hg.1 NM_032387.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700157726.hg.1 TC1700007915.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700157726.hg.1 ENST00000246914.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700157726.hg.1 ENST00000591448.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700157726.hg.1 NM_032387.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline