Search
Search

| Gene Symbol: H2AFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017991.2 | NP_001017991.1 | 1 | 148 | 117 | |
| GenBank mRNA | HY000925.1 | - | 1 | 149 | 117 | |
| Gene Symbol: H2AFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC130510.1 | - | 1 | 143 | 40146541 | 117 |
| BC130512.1 | - | 1 | 143 | 40146543 | 117 | |
Set Membership: |
|