| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000466599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000466599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000466599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029158.hg.1 |
ENST00000378509.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029158.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029158.hg.1 |
TC07001880.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000466599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000466599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000466599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012680.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
ENST00000378509.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
ENST00000416907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
ENST00000470969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
NM_032826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191305.hg.1 |
TC0700012680.hg.1 |
Clariom D TC |