Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00960523_m1
          See other SUFU GE Assays ›
          Gene Symbol:
          SUFU
          Gene Name:
          SUFU negative regulator of hedgehog signaling
          Gene Aliases:
          PRO1280, SUFUH, SUFUXL
          Chromosome Location:
          Chr.10: 102502879 - 102633458 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00960523_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SUFU
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001178133.1 NP_001171604.1 6 - 7 945 91
          NM_016169.3 NP_057253.2 6 - 7 945 91
          XM_011539860.2 XP_011538162.1 6 - 7 945 91
          GenBank mRNA AF144231.1 - 6 - 7 900 91
          AF159447.1 - 6 - 7 900 91
          AF172319.1 - 6 - 7 750 91
          AF175770.1 - 6 - 7 920 91
          AF222345.1 - 6 - 7 828 91
          AK312319.1 - 6 - 7 935 91
          AY358550.1 - 6 - 7 900 91
          BC013291.2 - 6 - 7 921 3533158 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008736.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000759.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SUFU

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          neural tube closure
          heart looping
          ventricular septum development
          regulation of transcription, DNA-templated
          proteolysis
          signal transduction
          multicellular organism development
          smoothened signaling pathway involved in ventral spinal cord interneuron specification
          smoothened signaling pathway involved in spinal cord motor neuron cell fate specification
          aorta development
          negative regulation of transcription factor import into nucleus
          cytoplasmic sequestering of transcription factor
          negative regulation of sequence-specific DNA binding transcription factor activity
          skin development
          negative regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          coronary vasculature development
          negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          transcription corepressor activity
          signal transducer activity
          protein binding
          beta-catenin binding
          transcription factor binding
          protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10009027.hg.1 ENST00000369899.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10009027.hg.1 ENST00000369902.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10009027.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10009027.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10009027.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10009027.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10009027.hg.1 TC10000759.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000159809.hg.1 TC1000008736.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369899.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000369902.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000423559.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 ENST00000471000.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_001178133.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008736.hg.1 NM_016169.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline