| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000478821.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000478821.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
ENST00000478821.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07012084.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07012084.hg.1 |
ENST00000420086.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07012084.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07012084.hg.1 |
TC07003342.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700198824.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700198824.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700198824.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700198824.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700198824.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700198824.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700198824.hg.1 |
TC0700013457.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000265825.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000421705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000469242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000478821.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000478821.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
ENST00000478821.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013457.hg.1 |
NM_020369.2 |
Refseq |