| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041427.hg.1 |
ENST00000295749.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041427.hg.1 |
ENST00000410053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041427.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041427.hg.1 |
TC02002582.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200210691.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200210691.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200210691.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200210691.hg.1 |
ENST00000404136.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200210691.hg.1 |
ENST00000410053.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200210691.hg.1 |
NM_020943.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200210691.hg.1 |
TC0200015099.hg.1 |
Clariom D TC |