| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015352.hg.1 |
ENST00000328853.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015352.hg.1 |
ENST00000381308.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015352.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015352.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015352.hg.1 |
TC19001117.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
TC1900009492.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176329.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000328853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000598081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000599020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
ENST00000620128.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_001244856.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009492.hg.1 |
NM_198492.3 |
Refseq |