Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00968940_m1
          See other NKX2-1 GE Assays ›
          Gene Symbol:
          NKX2-1
          Gene Name:
          NK2 homeobox 1
          Gene Aliases:
          BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1
          Chromosome Location:
          Chr.14: 36516397 - 36520232 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00968940_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NKX2-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079668.3 NP_001073136.1 2 - 3 566 72
          NM_003317.4 NP_003308.1 1 - 2 572 72
          GenBank mRNA BC006221.2 - 2 - 3 493 72
          BE796718.1 - 2 - 3 474 72
          BT009773.1 - 2 - 3 463 72
          D50740.1 - 3 - 4 617 72
          U19756.1 - 1 - 2 373 72
          U33749.1 - 1 - 2 714 72
          U43203.1 - 2 - 3 545 72
          X82850.1 - 1 - 2 387 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400010734.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14002321.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NKX2-1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neuron migration
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          phospholipid metabolic process
          pattern specification process
          axon guidance
          brain development
          endoderm development
          locomotory behavior
          response to hormone
          organ morphogenesis
          gene expression
          positive regulation of gene expression
          negative regulation of epithelial to mesenchymal transition
          telencephalon development
          globus pallidus development
          hippocampus development
          cerebral cortex cell migration
          forebrain dorsal/ventral pattern formation
          hypothalamus development
          forebrain neuron fate commitment
          forebrain neuron differentiation
          cerebral cortex GABAergic interneuron differentiation
          cerebral cortex neuron differentiation
          pituitary gland development
          telencephalon cell migration
          cell differentiation
          lung development
          negative regulation of cell migration
          negative regulation of transforming growth factor beta receptor signaling pathway
          thyroid gland development
          forebrain development
          developmental induction
          Leydig cell differentiation
          positive regulation of circadian rhythm
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          animal organ development
          anatomical structure formation involved in morphogenesis
          neuron fate commitment
          oligodendrocyte differentiation
          lung saccule development
          epithelial tube branching involved in lung morphogenesis
          Clara cell differentiation
          Type II pneumocyte differentiation
          GABAergic neuron differentiation
          interneuron migration
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          intronic transcription regulatory region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          enzyme binding
          sequence-specific DNA binding
          RNA polymerase II sequence-specific DNA binding transcription factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011990.hg.1 ENST00000498187.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011990.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011990.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011990.hg.1 TC14002321.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR14011989.hg.1 ENST00000498187.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011989.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011989.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011989.hg.1 TC14002321.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR14011988.hg.1 ENST00000498187.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011988.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011988.hg.1 TC14002321.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR14011991.hg.1 ENST00000498187.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011991.hg.1 NM_003317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011991.hg.1 TC14002321.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400182190.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182190.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182190.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182190.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182190.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182190.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182190.hg.1 TC1400010734.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000354822.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000518149.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_001079668.2 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010734.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182189.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182189.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182189.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182189.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182189.hg.1 TC1400010734.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400182192.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182192.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182192.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182192.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182192.hg.1 TC1400010734.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 ENST00000498187.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 ENST00000522719.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 ENST00000546983.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 NM_003317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400182191.hg.1 TC1400010734.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline