Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00969291_m1
          See other BNIP3 GE Assays ›
          Gene Symbol:
          BNIP3
          Gene Name:
          BCL2 interacting protein 3
          Gene Aliases:
          NIP3
          Chromosome Location:
          Chr.10: 131967683 - 131982013 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00969291_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BNIP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004052.3 NP_004043.3 3 - 4 496 125
          GenBank mRNA AF002697.1 - 3 - 4 415 125
          AK222626.1 - 3 - 4 406 125
          AK295136.1 - 3 - 4 406 125
          BC009342.2 - 3 - 4 391 125
          BC021989.1 - 3 - 4 410 4066029 125
          BC067818.1 - 3 - 4 439 125
          BC080643.1 - 3 - 4 391 125
          U15174.1 - 3 - 4 360 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000012329.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001769.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Recommended Assay Homo sapiens Human Apoptosis Human Antioxident Mechanisms Inventoried Human_Apoptosis Human_Antioxident_Mechanisms Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BNIP3

          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy
          response to hypoxia
          apoptotic process
          cell death
          granzyme-mediated apoptotic signaling pathway
          positive regulation of autophagy
          negative regulation of mitochondrial fusion
          positive regulation of cardiac muscle cell apoptotic process
          positive regulation of macroautophagy
          mitochondrial protein catabolic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of programmed cell death
          positive regulation of protein complex disassembly
          mitochondrial fragmentation involved in apoptotic process
          negative regulation of membrane potential
          regulation of mitochondrial membrane permeability
          autophagic cell death
          brown fat cell differentiation
          neuron apoptotic process
          defense response to virus
          response to hyperoxia
          negative regulation of cell death
          cellular response to hydrogen peroxide
          cellular response to mechanical stimulus
          cellular response to cobalt ion
          cellular response to hypoxia
          reactive oxygen species metabolic process
          positive regulation of mitochondrial fission
          positive regulation of release of cytochrome c from mitochondria
          mitochondrial outer membrane permeabilization
          toxin transport
          regulation of aerobic respiration
          intrinsic apoptotic signaling pathway in response to hypoxia
          negative regulation of reactive oxygen species metabolic process
          protein binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007721.hg.1 ENST00000605372.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007721.hg.1 ENST00000605372.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007721.hg.1 ENST00000605372.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10023342.hg.1 ENST00000368636.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10023342.hg.1 ENST00000540159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10023342.hg.1 NM_004052.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10023342.hg.1 TC10001769.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900010174.hg.1 ENST00000604515.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010174.hg.1 ENST00000604515.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010174.hg.1 ENST00000604515.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010174.hg.1 ENST00000604515.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000631806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay TC1000012329.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000184065.hg.1 TC1000012329.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200007721.hg.1 ENST00000605372.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007721.hg.1 ENST00000605372.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007721.hg.1 ENST00000605372.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 ENST00000368636.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 ENST00000540159.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 ENST00000633835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 NM_004052.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000184066.hg.1 TC1000012329.hg.1 Clariom D TC

          Back To Top
           

          Citations (1)

          Transglutaminase 2 protects against ischemic insult, interacts with HIF1beta, and attenuates HIF1 signaling.
          Authors
          Filiano AJ, Bailey CD, Tucholski J, Gundemir S, Johnson GV
          Journal
          Faseb J - Page No 2662 (Fri Aug 01 00:00:00 UTC 2008)
          Volume (Year)
          22 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline