| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000421208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07007121.hg.1 |
ENST00000450028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07007121.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07007121.hg.1 |
TC07000501.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000421208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008159.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
ENST00000414782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
ENST00000428436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
ENST00000450028.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
NR_003680.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156685.hg.1 |
TC0700008159.hg.1 |
Clariom D TC |