Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00970129_g1
          See other ALG12 GE Assays ›
          Gene Symbol:
          ALG12
          Gene Name:
          ALG12, alpha-1,6-mannosyltransferase
          Gene Aliases:
          CDG1G, ECM39, PP14673, hALG12
          Chromosome Location:
          Chr.22: 49860163 - 49918469 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00970129_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ALG12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024105.3 NP_077010.1 4 - 5 742 107
          XM_011530371.2 XP_011528673.1 4 - 5 753 107
          XM_017028936.1 XP_016884425.1 4 - 5 753 107
          XM_017028937.1 XP_016884426.1 4 - 5 753 107
          GenBank mRNA AF318343.1 - 4 - 5 694 107
          AJ290427.1 - 4 - 5 615 107
          AJ303120.1 - 4 - 5 615 107
          BC001729.1 - 4 - 5 589 3353085 107
          BC098562.1 - 4 - 5 635 30560020 107
          CR456369.1 - 3 - 4 531 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200009128.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000904.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ALG12

          Panther Classification:

          Molecular Function -

          glycosyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          protein N-linked glycosylation
          dolichol-linked oligosaccharide biosynthetic process
          mannosylation
          alpha-1,6-mannosyltransferase activity
          mannosyltransferase activity
          dol-P-Man:Man(7)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22015110.hg.1 ENST00000330817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22015110.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22015110.hg.1 TC22000904.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200165967.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2200165967.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200165967.hg.1 NM_024105.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200165967.hg.1 TC2200009128.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200165967.hg.1 ENST00000330817.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2200165967.hg.1 ENST00000492791.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200165967.hg.1 NM_024105.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline