Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00971099_m1
          See other MLLT3 GE Assays ›
          Gene Symbol:
          MLLT3
          Gene Name:
          MLLT3, super elongation complex subunit
          Gene Aliases:
          AF9, YEATS3
          Chromosome Location:
          Chr.9: 20341665 - 20622543 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00971099_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MLLT3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286691.1 NP_001273620.1 9 - 10 1698 144
          NM_004529.3 NP_004520.2 9 - 10 1782 144
          GenBank mRNA AK225957.1 - 9 - 10 1714 144
          AK297587.1 - 8 - 9 1809 144
          AK301474.1 - 9 - 10 1584 144
          AK312914.1 - 9 - 10 1675 144
          BC036089.1 - 9 - 10 1745 5298142 144
          BX649194.1 - 9 - 10 1758 144
          D16688.1 - 5 - 6 818 144
          KU178189.1 - 8 - 9 1495 144
          KU178190.1 - 4 - 5 386 144
          L13744.1 - 9 - 10 1690 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900009673.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000941.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MLLT3

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          segment specification
          anterior/posterior pattern specification
          negative regulation of canonical Wnt signaling pathway
          positive regulation of Wnt signaling pathway, planar cell polarity pathway
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000380338.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000468513.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000468513.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000468513.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000468513.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000468513.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000468513.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000469261.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000469261.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000469261.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000469261.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000469261.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000469261.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000475957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000475957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000475957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000475957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000475957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000475957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000475957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000488705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000488705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000488705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000488705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000488705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000488705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000491137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000491137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000491137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000491137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000491137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000491137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000629733.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 ENST00000630269.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_001286691.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009673.hg.1 NM_004529.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline