| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019776.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019776.hg.1 |
ENST00000499666.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019776.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019776.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019776.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019776.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019776.hg.1 |
TC04001416.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178164.hg.1 |
TC0400011421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000226522.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000499666.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
ENST00000515100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_001243736.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NM_021970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011421.hg.1 |
NR_024170.1 |
Refseq |