| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011867.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011867.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011867.hg.1 |
TC16000821.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000466154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
ENST00000486524.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009199.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170227.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170227.hg.1 |
NM_032444.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170227.hg.1 |
TC1600009199.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170227.hg.1 |
ENST00000294008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170227.hg.1 |
NM_032444.3 |
Refseq |