Search
Search
| Gene Symbol: API5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142930.1 | NP_001136402.1 | 1 - 2 | 240 | 129 | |
| NM_001243747.1 | NP_001230676.1 | 1 - 2 | 240 | 129 | ||
| NM_006595.3 | NP_006586.1 | 1 - 2 | 240 | 129 | ||
| XM_006718359.3 | XP_006718422.1 | 1 - 2 | 230 | 129 | ||
| GenBank mRNA | AF229254.1 | - | 1 - 2 | 180 | 129 | |
| AK293303.1 | - | 1 - 2 | 141 | 129 | ||
| AK299288.1 | - | 1 - 2 | 206 | 129 | ||
| AY311389.1 | - | 1 - 2 | 179 | 129 | ||
| BC017709.1 | - | 1 - 2 | 160 | 4419401 | 129 | |
| BM193468.1 | - | 1 - 2 | 222 | 129 | ||
| BT007093.1 | - | 1 - 2 | 67 | 129 | ||
| DC334531.1 | - | 1 - 2 | 240 | 129 | ||
| U83857.1 | - | 1 - 2 | 162 | 129 | ||
| Y15906.1 | - | 1 - 2 | 199 | 129 | ||
Set Membership: |
|