| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027784.hg.1 |
ENST00000239367.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027784.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027784.hg.1 |
ENST00000546019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027784.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027784.hg.1 |
TC06002217.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027785.hg.1 |
ENST00000239367.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027785.hg.1 |
ENST00000546019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027785.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027785.hg.1 |
TC06002217.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600202842.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202842.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202842.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202842.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202842.hg.1 |
TC0600014357.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202841.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202841.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202841.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202841.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202841.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202841.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202841.hg.1 |
TC0600014357.hg.1 |
Clariom D TC |