| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01032844.hg.1 |
ENST00000378251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01032844.hg.1 |
ENST00000462356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01032844.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01032844.hg.1 |
TC01002136.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
ENST00000479239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012601.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100193271.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193271.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193271.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100193271.hg.1 |
ENST00000378251.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193271.hg.1 |
ENST00000462356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193271.hg.1 |
NM_020710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100193271.hg.1 |
TC0100012601.hg.1 |
Clariom D TC |