Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00976237_m1
          See other MSTN GE Assays ›
          Gene Symbol:
          MSTN
          Gene Name:
          myostatin
          Gene Aliases:
          GDF8, MSLHP
          Chromosome Location:
          Chr.2: 190055700 - 190062729 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00976237_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MSTN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005259.2 NP_005250.1 1 - 2 506 69
          GenBank mRNA AF019627.1 - 1 - 2 373 69
          AF104922.1 - 1 - 2 506 69
          AK291524.1 - 1 - 2 477 69
          BC074757.2 - 1 - 2 420 30915216 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015221.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002618.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MSTN

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          transforming growth factor beta receptor signaling pathway
          muscle organ development
          response to heat
          response to gravity
          positive regulation of pathway-restricted SMAD protein phosphorylation
          skeletal muscle atrophy
          negative regulation of muscle hypertrophy
          response to muscle activity
          ovulation cycle process
          response to testosterone
          negative regulation of kinase activity
          regulation of apoptotic process
          skeletal muscle tissue regeneration
          regulation of MAPK cascade
          response to estrogen
          response to ethanol
          negative regulation of myoblast differentiation
          positive regulation of transcription, DNA-templated
          negative regulation of insulin receptor signaling pathway
          muscle cell cellular homeostasis
          cell development
          negative regulation of skeletal muscle tissue growth
          response to electrical stimulus
          negative regulation of protein kinase B signaling
          SMAD protein signal transduction
          cellular response to dexamethasone stimulus
          mitophagy in response to mitochondrial depolarization
          xenophagy
          negative regulation of skeletal muscle satellite cell proliferation
          negative regulation of satellite cell differentiation
          negative regulation of myoblast proliferation
          receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          heparin binding
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02041925.hg.1 ENST00000260950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041925.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041925.hg.1 TC02002618.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02041924.hg.1 ENST00000260950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041924.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041924.hg.1 TC02002618.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02041926.hg.1 ENST00000260950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041926.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041926.hg.1 TC02002618.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay TC0200015221.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211503.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211503.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211503.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211503.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211503.hg.1 TC0200015221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200211505.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211505.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211505.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211505.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211505.hg.1 TC0200015221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200211504.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211504.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211504.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211504.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211504.hg.1 TC0200015221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200211506.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211506.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211506.hg.1 ENST00000260950.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200211506.hg.1 NM_005259.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200211506.hg.1 TC0200015221.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline