Search
Search

OXNAD1| Gene Symbol: OXNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534235.2 | XP_011532537.1 | 9 - 10 | 1454 | 68 | |
| XM_017007490.1 | XP_016862979.1 | 9 - 10 | 1469 | 68 | ||
| XM_017007495.1 | XP_016862984.1 | 10 - 11 | 1596 | 68 | ||
Set Membership: |
|