Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00978218_m1
          See other KIF14 GE Assays ›
          Gene Symbol:
          KIF14
          Gene Name:
          kinesin family member 14
          Gene Aliases:
          MKS12
          Chromosome Location:
          Chr.1: 200551497 - 200620791 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00978218_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KIF14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001305792.1 NP_001292721.1 11 - 12 2387 162
          NM_014875.2 NP_055690.1 13 - 14 2799 162
          XM_011510231.2 XP_011508533.1 14 - 15 2893 162
          XM_011510232.2 XP_011508534.1 13 - 14 2555 162
          XM_011510233.2 XP_011508535.1 14 - 15 2686 162
          XM_011510235.2 XP_011508537.1 14 - 15 2398 162
          XM_011510236.1 XP_011508538.1 12 - 13 2682 162
          XM_017003005.1 XP_016858494.1 12 - 13 2976 162
          XM_017003006.1 XP_016858495.1 13 - 14 2584 162
          XM_017003007.1 XP_016858496.1 13 - 14 2111 162
          GenBank mRNA BC098582.1 - 3 - 4 392 162
          BC113742.1 - 12 - 13 2438 8322794 162
          D26361.1 - 13 - 14 2799 162

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100016891.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003679.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KIF14

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          microtubule-based movement
          microtubule depolymerization
          mitotic metaphase plate congression
          positive regulation of cell proliferation
          regulation of G2/M transition of mitotic cell cycle
          cerebellar granular layer structural organization
          cerebellar Purkinje cell layer structural organization
          cerebellar cortex development
          hippocampus development
          olfactory bulb development
          cell proliferation in forebrain
          cerebral cortex development
          regulation of cell adhesion
          regulation of cell migration
          cytoskeleton-dependent intracellular transport
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          regulation of myelination
          activation of protein kinase activity
          positive regulation of cytokinesis
          regulation of Rap protein signal transduction
          negative regulation of integrin activation
          substrate adhesion-dependent cell spreading
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          establishment of protein localization
          cell division
          response to docetaxel trihydrate
          mitotic cell cycle process
          regulation of cell maturation
          regulation of G1/S transition of mitotic cell cycle
          microtubule motor activity
          protein binding
          ATP binding
          microtubule binding
          ATP-dependent microtubule motor activity, plus-end-directed
          tubulin binding
          ATPase activity
          protein kinase binding
          PDZ domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01056336.hg.1 ENST00000367350.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01056336.hg.1 NM_014875.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01056336.hg.1 TC01003679.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 ENST00000367350.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 ENST00000614960.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 NM_001305792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 NM_014875.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 ENST00000367350.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 ENST00000614960.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 NM_001305792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 NM_014875.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100227716.hg.1 TC0100016891.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline