Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00978328_m1
          See other SIRT5 GE Assays ›
          Gene Symbol:
          SIRT5
          Gene Name:
          sirtuin 5
          Gene Aliases:
          SIR2L5
          Chromosome Location:
          Chr.6: 13574222 - 13615158 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00978328_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SIRT5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012241.4 NP_036373.1 1 - 2 119 153
          GenBank mRNA BC000126.1 - 1 - 2 103 3352592 153
          DC314190.1 - 1 - 2 119 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006992.hg
          This assay targets the same location as HTA transcript cluster(s) TC06000093.hg

          Additional Information:

          This assay is in the 5'UTR region of the BC000126.1,NM_012241.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SIRT5

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin silencing
          protein ADP-ribosylation
          protein deacetylation
          regulation of ketone biosynthetic process
          negative regulation of cardiac muscle cell apoptotic process
          response to nutrient levels
          protein demalonylation
          peptidyl-lysine demalonylation
          protein desuccinylation
          peptidyl-lysine desuccinylation
          protein deglutarylation
          peptidyl-lysine deglutarylation
          negative regulation of reactive oxygen species metabolic process
          NAD+ ADP-ribosyltransferase activity
          zinc ion binding
          protein-malonyllysine demalonylase activity
          protein-succinyllysine desuccinylase activity
          protein-glutaryllysine deglutarylase activity
          NAD+ binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000359782.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000379262.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000397350.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606427.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606427.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606427.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606427.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 ENST00000606427.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001193267.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_001242827.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_012241.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006992.hg.1 NM_031244.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 146299 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline